Goggles, DuraMaxx™
Anti-UV, antigraffio, antiappannamento; con fascia in neoprene, In PC grigio TSR, blu/grigio
Codice catalogo 111-1433
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
These low profile goggles with toric lens provide superior optics and unobstructed vision, while the soft, elastomer bodies flex and conform to the face for a comfortable, gap-free fit.
- PP/TPE frame and large adjustable headband
- Indirect venting system minimises fogging to keep lenses clear
- Dura-streme™ lens coating provides anti-scratch coating on the outside and anti-fog coating on the inside
The OTG goggles fit over most prescription spectacles comfortably. Clear lens model is adapted for indoor use, grey model for outdoor use.
Weight: 100 g
Certifications: EN 166
Specifiche
- Tipo:Anti-UV, antigraffio, antiappannamento; con fascia in neoprene
- Colore:blu/grigio
- Seq:4
- Materiale lente:In PC grigio TSR