Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 7950 series, width 65 mm, BBP™11, 70 m, nero
BRDY804467
: Brady
Documents
- Colore:nero
- Descrizione:Thermal transfer printer ribbon, 7950 series, width 65 mm
- Lunghezza:70 m
- Seq:02
- Per:BBP™11