Cover slides
24×50 mm, 0,15 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Clear and pre-washed glass cover slides dedicated to sample and lens protection.
Delivery information: Supplied in cases of one or two hundred.
Documents
Certificates
Search by Lot NumberSpecifications
- L×L:24×50 mm
- Seq:02
- Spessore:0,15 mm