Jars with lid
250 ml, 79,5 mm, 84,5 mm, 62 mm
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Jars, made of PP, translucent, with lid.
- Suitable for ailmentary use
- Supplied uncapped
- Good chemical and temperature resistance
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Ø est:79,5 mm
- Capacità:250 ml
- Altezza:62 mm
- Seq:01
- Ø interno collo:84,5 mm