Petri dishes
Vented, with 2 compartments, 90 mm, 14 mm, Asettico, 25 bags of 20
Katalognr. DELT200202
Anbieter: DELTALAB
Direkt zu
Product Details & Documents
Vented petri dishes made from PS.
Technische Daten
- Ø:90 mm
- Confezione:25 bags of 20
- Sterilità:Asettico
- Versione:Vented, with 2 compartments
- Altezza:14 mm
- Seq:04