Thermal transfer printer labels, type B-423
Printer labels, B-423, THT-18-423-3, Poliestere, Acrilico permanente, Thermal transfer, bianco, 360X-Plus II, 76×25 mm, 3000 labels/roll
Codice catalogo BRDY030134
Nome del fornitore: Brady
Thermal transfer printer labels, type B-423
Codice catalogo BRDY030134
Nome del fornitore: Brady
Vai a
Product Details & Documents
Polyester labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves
This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
TDS B-423 Labels and Signs TDS B-423Y Labels and Signs TDS Bradybondz B-423 Thermal Transfer Printable Glossy White Polyester Label Stock TDS Bradybondz B-423Y Thermal Transfer Printable Glossy Yellow Polyester Label Stock TDS Bradybondz™ B-423 thermal transfer printable glossy white polyester label stock
Specifiche
- Stampante:360X-Plus II
- Printing method:Thermal transfer
- Tipo:B-423
- Colore:bianco
- Descrizione:Printer labels
- Dimensioni:76×25 mm
- Materiale:Poliestere
- Seq:19
- Modello:THT-18-423-3
- Confezione:3000 labels/roll
- Adesivo:Acrilico permanente



