Provette per centrifuga
50 ml, 29×118,5 mm, Tappo a vite, 7000×g, Skirted, Sterile, 100 per bag/500 per case
Supplier: DELTALAB
Jump to:
Product Details & Documents
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
Specifications
- Tipo di chiusura:Tappo a vite
- Ø×L:29×118,5 mm
- Confezione:100 per bag/500 per case
- Tipo:Skirted
- Versione:Sterile
- Capacità:50 ml
- Seq:07
- RCF max.:7000×g