Provette per centrifuga
15 ml, 17×122 mm, Tappo a vite, 5000×g, Without skirt, Sterile, Individually bagged
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Tubes made of PP, translucent, with blue HDPE screw cap.
- Continuous thread
- External moulded graduations in 0,5 ml increments
- Excellent temperature resistance
- Excellent chemical resistance
Suitable for centrifugation tests in immunology, microbiology, more.
Specifiche
- Tipo di chiusura:Tappo a vite
- Ø×L:17×122 mm
- Confezione:Individually bagged
- Tipo:Without skirt
- Versione:Sterile
- Capacità:15 ml
- Seq:03
- RCF max.:5000×g