Bandage mull
Gauze, 20-ply, zigzag layers, 80 cm wide, 10 m long
Codice catalogo HART206406/3
Nome del fornitore: HARTMANN PAUL
Vai a
Product Details & Documents
Mull for making individual bandages and wound dressings.
- 20 threads
- Laid 8-fold in zigzag layers to a width of 10 cm
- Good purity and absorbency
Certifications: EN 14079
Specifiche
- Descrizione:Gauze, 20-ply, zigzag layers, 80 cm wide, 10 m long