Nessler cylinders
Nessler cylinder 100 ml, low form, with optically plane bottom, circular mark at 100 ml, Ø×L: 34×180 mm
Codice catalogo WITG2.151.100
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Specifiche
- Descrizione:Nessler cylinder 100 ml, low form, with optically plane bottom, circular mark at 100 ml, Ø×L: 34×180 mm
- Seq:05