Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Chemo protective aprons, BioClean-C™

Chemo protective aprons, BioClean-C™

blu, S, +
Codice catalogo 113-8167
Nome del fornitore:   Nitritex
Chemo protective aprons, BioClean-C™
Codice catalogo 113-8167
Nome del fornitore:  Nitritex
Chemo protective aprons, BioClean-C™
Chemo protective aprons, BioClean-C™

blu, S, +

Price
512.00 SFr.
Log in per disponibilità
1 * 50 ST
Product Details & Documents

These disposable aprons are made of CleanTough™ fabric, a spunbonded non woven PP fabric laminated with a PE film.

  • Processed to ensure ISO Class 4 compatibility
  • Lightweight, low linting and hard-wearing CleanTough™ fabric
  • Adjustable neck fastening
  • Tie tapes at the waist

The aprons provide protection against fine sprays and particles as well as against a range of chemotherapy drugs.

Colour: Blue

Certifications: EN ISO 13034:2005 +A1:2010 (Type PB [6]), EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.

Packaging: One piece per sealed inner bag; one sealed inner bag per sealed outer bag; 50 outer bags per carton liner; one carton liner per carton (50 pieces).

Documents

Specifiche
  • Taglia dell'abbigliamento:
    S
  • Sterili:
    +
  • Colore:
    blu
  • Seq:
    04
  • Cat. No.:
    113-8167
  • Applicazioni per abiti:
    Medical|Clinical