Counting Chambers, Neubauer
Counting chamber with double ruling, Neubauer, 0,02 mm
Codice catalogo HECH92044102702
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Counting Chambers, Neubauer
Codice catalogo HECH92044102702
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
These counting chambers are designed for blood tests and urine analysis.
- Counting erythrocytes corresponds exactly to Thoma method
- Counting leucocytes in five large squares
- Evaluation just as indicated for Thoma ruling
Consisting of nine large squares – The central square shows the Thoma ruling and the other eight squares are subdivided.
Documents
HECH40441702_TDS_EN
Datasheet
Specifiche
- Profondità della camera:0,02 mm
- Descrizione:Counting chamber with double ruling, Neubauer
- Seq:03