Nitrile gloves, Omega™ Pro-Tech
XS, porpora, 240 mm, 10 boxes, 100 each
NITROPT2101-XS
: Nitritex
These nitrile gloves feature an ambidextrous shape and beaded cuffs. They are resistant to a wide range of chemicals.
- Non sterile, powder-free
- Contain no natural latex proteins
- Textured for improved grip
- Beaded cuff provides easier donning and stability on the arm
- Each size carries a unique bar code on the inner dispenser box (EAN-13) and the outer carton (ITF-14), which allows automatic data entry into stock control systems
: EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010; Cat. III; AQL 1,5
Food contact approved
- Dimensione del guanto:XS
- Confezione:10 boxes, 100 each
- Colore:porpora
- Lunghezza:240 mm
- Seq:01
- Monouso/riutilizzabile:monouso
- Talcati/non talcati:Non talcati
- Norma EN:EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010
- Cat. No.:NITROPT2101-XS
- Ambidestro/mano specifico:ambidestro
- Lattice/senza lattice:Senza lattice
- Approvati per il contatto con gli alimenti:+
- Modello:Pro-Tech
- Finitura:Textured
- Tipo polsino:Cuff_Style-21391103|Cuff_Style-21391099