Burner, Teclu
Teclu burner with air regulation needle valve, propane gas, head Ø 32 mm, tubing Ø 19 mm, Propano, 32 mm
Catalog # USBE1301
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Nickel plated brass base, in powder coated grey.
- With air regulation valve and throttle screw
- Without pilot light
Height: 165 mm
Specifications
- Descrizione:Teclu burner with air regulation needle valve, propane gas, head Ø 32 mm, tubing Ø 19 mm
- Ø testa:32 mm
- Per:Propano