Glass Microfiber Filter Paper
GF/C, 27 mm
Codice catalogo PRATA0908855
Nome del fornitore: PRAT DUMAS
Vai a
Product Details & Documents
These glass microfiber filter papers allow fast filtration and high particle retention. They can be used at temperatures up to 380 °C.
- Chemical compatibility with many solvents and with reagents
- Suitable for microfiltration
Specifiche
- Grado:GF/C
- Seq:2
- Taglia:27 mm
- Cat. No.:PRATA0908855
- Peso:55 g/m²
- Spessore:270 µm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



