Orders LinkContactUsLinkComponent
PCR tubes, 0,2 ml, with cap
Individual tube with flat cap, naturale, 0,2 ml
  DELT4094.1N
 :   DELTALAB
PCR tubes, 0,2 ml, with cap
  DELT4094.1N
 :  DELTALAB
PCR tubes, 0,2 ml, with cap
Individual tube with flat cap, naturale, 0,2 ml
SFr. 172.00
1 * 1.000 ST
Tubes made of PP with attached hinged caps.
  • Caps are flat and easily pierceable
  • RNAse, DNAse and PCR inhibitors free
  • Capacità:
    0,2 ml
  • Colore:
    naturale
  • Descrizione:
    Individual tube with flat cap
  • Seq:
    01