Filling and storage solution for combined/reference electrodes
Filling and storage solution for combined/reference electrodes, KCl 3M (without AgCl), 500 ml bottle, 500 mL
Codice catalogo HACHC20C320
Nome del fornitore: Hach
Filling and storage solution for combined/reference electrodes
Codice catalogo HACHC20C320
Nome del fornitore: Hach
Numero CAS:
Vai a
Product Details & Documents
Shelf life: 48 months from production date.
Specifiche
Specifiche
- Conf:500 mL
- Descrizione:Filling and storage solution for combined/reference electrodes, KCl 3M (without AgCl), 500 ml bottle