Orders LinkContactUsLinkComponent
PCR tubes, 0,2 ml, with cap
PCR tubes, 0,2 ml, with cap
Individual tube with flat cap, rosso, 0,2 ml
Codice catalogo DELT4094.1R
Nome del fornitore:   DELTALAB
PCR tubes, 0,2 ml, with cap
Codice catalogo DELT4094.1R
Nome del fornitore:  DELTALAB
PCR tubes, 0,2 ml, with cap
Individual tube with flat cap, rosso, 0,2 ml
Price
SFr. 207.00
Log in per disponibilità
1 * 1.000 ST
Product Details & Documents
Tubes made of PP with attached hinged caps.
  • Caps are flat and easily pierceable
  • RNAse, DNAse and PCR inhibitors free
Documents
Specifiche
  • Capacità:
    0,2 ml
  • Colore:
    rosso
  • Descrizione:
    Individual tube with flat cap
  • Seq:
    02