PCR tubes, 0,2 ml, with cap
Individual tube with flat cap, rosso, 0,2 ml
Codice catalogo DELT4094.1R
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Tubes made of PP with attached hinged caps.
- Caps are flat and easily pierceable
- RNAse, DNAse and PCR inhibitors free
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Capacità:0,2 ml
- Colore:rosso
- Descrizione:Individual tube with flat cap
- Seq:02