Adapters with external thread
Adapter N with 0,7 mm O.Ø Pt/lr capillary nipple
Codice catalogo PERCB0507949
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
Adapters with external screw thread (6,4 mm) for FIMS-100, FIMS-400, FIAS-100 or FIAS-400 flow injection system.
Specifiche
- Descrizione:Adapter N with 0,7 mm O.Ø Pt/lr capillary nipple
- Seq:04