Adapters with external thread
Adapter L with 2,8 mm O.Ø nipple
Codice catalogo PERCB0507920
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
Adapters with external screw thread (6,4 mm) for FIMS-100, FIMS-400, FIAS-100 or FIAS-400 flow injection system.
Specifiche
- Descrizione:Adapter L with 2,8 mm O.Ø nipple
- Seq:02