Closures for headspace vials
Blue PTFE coated silicone Bi-metal magnetic cap, pre-assembled
Codice catalogo PERCN6356566
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifiche
- Descrizione:Blue PTFE coated silicone Bi-metal magnetic cap, pre-assembled
- Seq:18