Microsampling tools
Dissecting forceps
Codice catalogo PERCN9302610
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Informazioni per l'ordine: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifiche
- Descrizione:Dissecting forceps
- Seq:02