Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Headspace screw top vials

Headspace screw top vials

Headspace screw top vials with write-on patch and fill lines, 20,00 ml, 23×75,5 mm, Vetro, trasparente, No
Codice catalogo PERCN9306241
Nome del fornitore:   PerkinElmer
Headspace screw top vials
Codice catalogo PERCN9306241
Nome del fornitore:  PerkinElmer
Headspace screw top vials

Headspace screw top vials with write-on patch and fill lines, 20,00 ml, 23×75,5 mm, Vetro, trasparente, No

Price
SFr. 1’290.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Tappo incluso:
    No
  • Ø×A:
    23×75,5 mm
  • Capacità:
    20,00 ml
  • Colore:
    trasparente
  • Descrizione:
    Headspace screw top vials with write-on patch and fill lines
  • Materiale:
    Vetro
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.