Closures for headspace vials
Red PTFE coated butyl cap/septa/spring
Catalog # PERCN9303991
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
- Descrizione:Red PTFE coated butyl cap/septa/spring
- Seq:25