Orders LinkContactUsLinkComponent
Interference check standards for ICP-OES
Interference check standards for ICP-OES
Soluzione di analiti C, 2 mg/L: Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg*, Mn, Ni, Pb, Sb, Se, Tl, V, Zn, 1
Catalog # PERCN9303831
Supplier:   PerkinElmer
Interference check standards for ICP-OES
Catalog # PERCN9303831
Supplier:  PerkinElmer
CAS Number:  
Interference check standards for ICP-OES
Soluzione di analiti C, 2 mg/L: Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg*, Mn, Ni, Pb, Sb, Se, Tl, V, Zn, 1
CAS Number:  
Price
SFr. 671.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
Specifications
  • Conf:
    1
  • Descrizione:
    Soluzione di analiti C
  • Indice:
    2 mg/L: Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg*, Mn, Ni, Pb, Sb, Se, Tl, V, Zn
Specifications