Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 1,0 mm thickness for semi-demountable cell, SpectrumGX
Codice catalogo PERCL1270410
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specifiche
- Per:SpectrumGX
- Descrizione:Spacers, PTFE, rectangular, 1,0 mm thickness for semi-demountable cell
- Seq:06