Orders LinkContactUsLinkComponent
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Single channel pipettes, mechanical, fixed / variable volume, labopette®
250 µl, ±0,8 %, <0,3 %, Blue, 10 - 1000 µl
Catalog # HIRS9475508
Supplier:   HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Catalog # HIRS9475508
Supplier:  HIRSCHMANN
Single channel pipettes, mechanical, fixed / variable volume, labopette®
Single channel pipettes, mechanical, fixed / variable volume, labopette®
250 µl, ±0,8 %, <0,3 %, Blue, 10 - 1000 µl
Price
310.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
The reliable labopette® pipette features superior ergonomic characteristics, right down to the smallest detail. The entire working process is optimised to ensure non fatiguing work that places a minimum of stress on the joints.
  • Volume configuration with digital locking feature
  • With tip ejection and calibration option
  • Tip seal and safety filter to prevent contamination
  • Trigger grip ensures a firm but relaxed hold during pipetting
  • Ergonomic design of the handle offers an equally firm, comfortable hold for large and small hands, right and left-handed users and hands with or without gloves
  • Effective thermal insulation of the unit interior prevents hand heat penetrating into the interior of the unit
  • Completely autoclavable without dismantling
Informations de livraison: Supplied with calibration tool, pipette holder, individual serial number and calibration certificate.
Specifications
  • Volume:
    250 µl
  • Seq:
    16
  • Accuratezza:
    ±0,8 %
  • Imprecisione:
    <0,3 %
  • Per puntali:
    Blue, 10 - 1000 µl
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470010
    Filtri Safe-Cone, per labopette®; volume variabile 1000 - 5000 µl; volume fisso 2000/5000 µl
    Log in for availability
    1 * 50 ST - SFr. 72.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470013
    Filtri Safe-Cone, per labopette® a volume variabile 2 - 20/5 - 50 µl; volume fisso 20/25/50 µl
    Log in for availability
    1 * 50 ST - SFr. 72.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470011
    Filtri Safe-Cone, per labopette® electronic 10 - 500/50 - 1000/500 - 5000/50 - 1200 µl; volume variabile 100 - 1000 µl; volume fisso 250/500/1000 µl
    Log in for availability
    1 * 50 ST - SFr. 72.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470012
    Filtri Safe-Cone, per labopette® a volume variabile 10 - 100/20 - 200 µl; volume fisso 100/200 µl
    Log in for availability
    1 * 50 ST - SFr. 72.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9980116
    Certificato di calibrazione DKD per labopette® a volume fisso 0,5 μl - 5 ml / labopettor
    Log in for availability
    1 * 1 ST - SFr. 102.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9980118
    Certificato di calibrazione DKD per labopette® a 12 canali, 5 - 250 μl
    Log in for availability
    1 * 1 ST - SFr. 449.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470006
    Supporto per sei pipette meccaniche
    Log in for availability
    1 * 1 ST - SFr. 107.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470004
    Supporto a carosello per sei pipette meccaniche
    Log in for availability
    1 * 1 ST - SFr. 217.00
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490101
    0,2 - 10 µl
    Trasparente
    Pipette Hirschmann Laborgeräte, Gilson®
    Log in for availability
    1 * 1.000 ST - SFr. 42.00
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490102
    1 - 200 µl
    Giallo
    Pipette Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson®
    Log in for availability
    1 * 1.000 ST - SFr. 28.10
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490103
    100 - 1000 µl
    Blu
    Pipette Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson®
    Log in for availability
    1 * 1.000 ST - SFr. 31.20
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490106
    1 - 250 µl
    Trasparente
    Pipette multicanale labopette®
    Log in for availability
    1 * 1.000 ST - SFr. 45.50
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490124
    100 - 5000 µl
    Trasparente
    Pipette Hirschmann Laborgeräte
    Log in for availability
    1 * 300 ST - SFr. 66.70
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490201
    0,2 - 10 µl
    Trasparente
    Pipette Hirschmann Laborgeräte, Gilson®
    Log in for availability
    1 * 960 ST - SFr. 101.00
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490202
    1 - 200 µl
    Giallo
    Pipette Hirschmann Laborgeräte, Eppendorf®, Brand®, Gilson®
    Log in for availability
    1 * 960 ST - SFr. 75.50
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490205
    50 - 1200 µl
    Trasparente
    Pipette Hirschmann Laborgeräte
    Log in for availability
    1 * 960 ST - SFr. 187.00
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490206
    1 - 250 µl
    Trasparente
    Pipette multicanale labopette®
    Log in for availability
    1 * 960 ST - SFr. 75.00
Product Information
  • Catalog #
    Volume
    Colore puntale
    Per uso con
    Availability
    Price
  • HIRS9490104
    1 - 10000 µl
    Trasparente
    Pipette Hirschmann Laborgeräte
    Log in for availability
    1 * 100 ST - SFr. 66.70
Product Information
  • Catalog #
    Stile punta
    Per uso con
    Availability
    Price
  • HIRS9490109
    Multibox, autoclavable
    refill trays of pipette tips up to 300 µl
    Log in for availability
    1 * 1 ST - SFr. 35.80
Product Information
  • Catalog #
    Stile punta
    Per uso con
    Availability
    Price
  • HIRS9490110
    Multibox, autoclavable
    refill trays of pipette tips up to 1000 µl
    Log in for availability
    1 * 1 ST - SFr. 37.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9470014
    Filtri Safe-Cone, per labopette® a volume fisso 10/20/25/250 µl; multicanale 5,0 - 50/50 - 300 µl; elettronica multicanale 25 - 250/5 - 100 µl
    Log in for availability
    1 * 50 ST - SFr. 72.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9980115
    Certificato di calibrazione DKD per labopette® a volume variabile 0,5 μl - 5 ml / labopettor
    Log in for availability
    1 * 1 ST - SFr. 166.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HIRS9980117
    Certificato di calibrazione DKD per labopette® a 8 canali, 0,5 - 250 μl
    Log in for availability
    1 * 1 ST - SFr. 413.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.