Multiple use earplugs, SmartFit®
Multiple use earplugs with cord, detectable, blu
Codice catalogo 111-0556
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
These earplugs with Conforming Material Technology™ use body heat to adapt to the shape of the ear canal for a comfortable, personalized fit.
- Three-flange design delivers superior comfort and a truly individual fit
- Detachable cord system
- Metal ring on stem detectable by automated equipment (SmartFit Detectable only)
The detectable version with its bright blue color and metalized features can be detected by visual or automated methods and is therefore ideal for food applications.
SNR: 25 dB
Certifications: EN 352-2
Informazioni per l'ordine: Supplied with HearPack® storage case.
Specifiche
- Colore:blu
- Descrizione:Multiple use earplugs with cord, detectable
- Seq:002