Degustation and drinking cups
100 ml, 75 mm
Codice catalogo SEMA5652
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Polystyrene, clear.
- Good chemical resistance
- Good temperature resistance
These dugustation and drinking cups are crystal clear, sprayed design, well standing, stable and elegant shape with content marking.
Specifiche
- Capacità:100 ml
- Altezza:75 mm
- Seq:02