Powder funnels, PP/PE
100 mm, 22 mm
Codice catalogo BURK9606-0100
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
These powder funnels are designed for safe filling of containers.
- PP/PE
9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Specifiche
- Ø esterno imbuto:100 mm
- Seq:02
- Ø esterno stelo:22 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



