Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
100 ml, 29/32 NS
Codice catalogo NEUB0752-29-0100
Nome del fornitore:   NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
Codice catalogo NEUB0752-29-0100
Nome del fornitore:  NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
100 ml, 29/32 NS
Price
SFr. 46.90
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Specifiche
  • Cono femmina:
    29/32 NS
  • Capacità:
    100 ml
  • Seq:
    04