Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Chlorophosphonazo indicator solution for hardness determination

Chlorophosphonazo indicator solution for hardness determination

Chlorophosphonazo indicator solution, 500 ml, 500 mL
Catalog # HACH2589549
Supplier:   Hach
Chlorophosphonazo indicator solution for hardness determination
Catalog # HACH2589549
Supplier:  Hach
CAS Number:  
Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution for hardness determination

Chlorophosphonazo indicator solution, 500 ml, 500 mL

CAS Number:  
Price
SFr. 1’830.00
Log in for availability
1 * 500 mL
Product Details & Documents

For the determination of ultra low range hardness by chlorophosphonazo method. Ideal for use with Pour-Thru cell. Requires CDTA reagent.

Specifications

Specifications

  • Conf:
    500 mL
  • Descrizione:
    Chlorophosphonazo indicator solution, 500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.