Tank liners, HyPerforma™
Tank liners, 50 l
Codice catalogo HYCPSH30646.01
Nome del fornitore: THERMO BIOPROCESS DIVISION
Tank liners, HyPerforma™
Codice catalogo HYCPSH30646.01
Nome del fornitore: THERMO BIOPROCESS DIVISION
Vai a
Product Details & Documents
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
- Gusset design allows for a close fit to the tank
- Constructed of CX3-9 film
- For HyPerforma™ Single-Use Mixer DS 300
Documents
Specifiche
- Descrizione:Tank liners, 50 l
- Seq:01