Storage Boxes with Lid
114×60×23 mm
SEMA2251
: SEMADENI
These storage boxes with lid, made of polystyrene (PS), are transparent and have a plastic hinged lid.
- Seq:02
- L×P×A:114×60×23 mm
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
#: 20864.320
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
#: 83640.320
: VWR Chemicals
Sodio idrossido 0.1 mol/l (0.1 N) in soluzione acquosa, AVS TITRINORM Reag. Ph. Eur. soluzione volumetrica
#: 31770.294
: VWR Chemicals