Wringers for mops
Stainless steel heavy-duty downpress wringer for VertiKlean® MAX™, QuickTask™ and Edgeless® mops, 380×125×100 mm
Codice catalogo 141-0959
Vai a
Discontinued
Product Details & Documents
Variety of wringers for curved and flat mops.
- Made of stainless steel
- Can be used with curved, flat, and string mops
- Ideal for use with Contec’s 15, 20, and 25 L bucket systems
- Fully autoclavable
Specifiche
- Descrizione:Stainless steel heavy-duty downpress wringer for VertiKlean® MAX™, QuickTask™ and Edgeless® mops
- Dimensioni:380×125×100 mm
- Seq:06