Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
24/29 NS, 24/29 NS, 150 mm
Codice catalogo 201-0598
Nome del fornitore:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Codice catalogo 201-0598
Nome del fornitore:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
24/29 NS, 24/29 NS, 150 mm
Price
115.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifiche
  • Dimensione cono:
    24/29 NS
  • Cono femmina:
    24/29 NS
  • Lunghezza totale:
    150 mm