Adapters, Combifix®
Luer-Lock, maschio/maschio, trasparente
Nome del fornitore: B. Braun
Vai a
Product Details & Documents
These adapters are used for connecting fittings.
Suitable for interconnecting male and female fittings with 6% (Luer) and 10% (Rekord) taper fittings.
Documents
Specifiche
- Colore:trasparente
- Descrizione:Luer-Lock, maschio/maschio
- Seq:03