Conductive pipette tips
Conductive, slim style, 10 - 240 µl, -, -, Nero, Bulk, 900/bag
Codice catalogo GREI764280_7200
Nome del fornitore: Greiner Bio-One
Vai a
Product Details & Documents
Conductive tips are used to prevent carryover in automated pipetting robots. The addition of graphite to the raw material PP makes the pipette tips electrically conductive. By measuring electric currents, the filling level can be determined in sample and reagent containers, so that the depth of immersion of the tip can be adjusted to the filling level.
- Filter tip: Free of detectable DNase, RNase, human DNA; non pyrogenic, non cytotoxic
Specifiche
- Confezione:Bulk, 900/bag
- Sterili:-
- Volume:10 - 240 µl
- Stile punta:Conductive, slim style
- Colore puntale:Nero
- Filtro:-