Powder funnels
65 mm
Codice catalogo HECH44002030
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
- Excellent chemical resistance
- Excellent temperature resistance
- Autoclavable
Documents
HECH44002044_TDS_EN
Datasheet
Specifiche
- Ø esterno imbuto:65 mm