Orders LinkContactUsLinkComponent
One-Step SYBR® Green qRT-PCR, qScript™
One-Step SYBR® Green qRT-PCR, qScript™
qScript™ One-Step SYBR® Green qRT-PCR Kit, 200×50 µl reactions, Agilent AriaMx, BioRad CFX, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays
Codice catalogo 733-2078
Nome del fornitore:   Quantabio
One-Step SYBR® Green qRT-PCR, qScript™
Codice catalogo 733-2078
Nome del fornitore:  Quantabio
Numero CAS:  
One-Step SYBR® Green qRT-PCR, qScript™
qScript™ One-Step SYBR® Green qRT-PCR Kit, 200×50 µl reactions, Agilent AriaMx, BioRad CFX, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays
Numero CAS:  
Price
SFr. 804.00
Log in per disponibilità
Product Details & Documents
qScript™ One-Step SYBR® Green qRT-PCR Kits reliably deliver maximum qRT-PCR efficiency, sensitivity and specificity in with a simplified protocol. These kits offer reliable and unbiased amplification of all RNA targets regardless of abundance. The proprietary supermix maximises activities of both reverse transcriptase and AccuStart™ Taq DNA polymerase without compromising specificity of amplification. qScript™ One-Step SYBR® Green qRT-PCR kits are available and optimised for all real-time PCR instrument platforms including those requiring normalisation with ROX reference dye.
Informazioni per l'ordine: Kits are available with or without ROX or with fluorescein for Bio-Rad iCycler iQ® systems. All kits contain qScript One-Step Reverse Transcriptase (optimised 50X formulation of recombinant MMLV reverse transcriptase for one-step RT-PCR); One-Step SYBR Green Master Mix (2X) reaction buffer containing dNTPs, MgCl₂, AccuStart™ Taq DNA polymerase, stabilisers, and SYBR® Green I dye; and nuclease-free water.
Specifiche
Specifiche
  • Compatibilità con il ciclatore:
    Agilent AriaMx, BioRad CFX, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
  • Conf:
    200 Assays
  • Descrizione:
    qScript™ One-Step SYBR® Green qRT-PCR Kit, 200×50 µl reactions
  • Cat. No.:
    733-2078
  • Applicazione:
    RT-qPCR dye
  • One step:
    Yes