SampliQ SPE cartridges
Cartuccia, Si, 60 Å, 45 µm, 3 ml, 500 mg
Codice catalogo 549-0587
Nome del fornitore: AGILENT HEWLETT PACKARD
Vai a
Product Details & Documents
High quality SampliQ SPE (Solid Phase Extraction) products help the user confidently extract and concentrate samples from complex matrices, ensuring fast, accurate, and reproducible results from the very first step. The SampliQ line includes polymers, silica and non-silica options to meet user's specific SPE needs.
- Improve and simplify separations
- Increase column lifetime
- Improve detection limits
SampliQ SPE products are manufactured in the US to ISO-9001 standards.
Confezione: To ensure product integrity all SampliQ products are packaged with triple lamination and include a certificate of performance.
Specifiche
- Formato della colonna:Cartuccia
- Dimensioni particelle:45 µm
- Dimensioni pori:60 Å
- Capacità:3 ml
- Seq:11
- Peso letto:500 mg
- Fase:Si