Maschera, Kimtech™ M7, PE
Con bottoni, bianco
Codice catalogo 113-0929
Nome del fornitore: Ansell
Vai a
Product Details & Documents
Single-use personal veil designed to fit most garment systems.
- Made from low linting breathable PE film
- Knitted headbands or snap fasteners
- Intended for use in cleanrooms where reusable face veils are currently worn
Suitable for cleanrooms ISO class 7 or higher.
Confezione: 25 veils per double bag, 6 bags per case.
Documents
Kimberly-Clark Kimtech™ Cleanroom Face Masks
Letteratura sul prodotto
Specifiche
- Tipo:Con bottoni
- Colore:bianco