Orders LinkContactUsLinkComponent
Vario couplings
Vario couplings
Couplings with UNF1/4"28G female thread, 1,6 mm, Connecting flanged PTFE
elastic tubing with end fittings and wall thickness 0,8 and 1,6 mm, 1/4“ 28G UNF, 1,6 mm
Catalog # BOHLF778-16
Supplier:   Bohlender
Vario couplings
Catalog # BOHLF778-16
Supplier:  Bohlender
Vario couplings
Vario couplings
Couplings with UNF1/4"28G female thread, 1,6 mm, Connecting flanged PTFE
elastic tubing with end fittings and wall thickness 0,8 and 1,6 mm, 1/4“ 28G UNF, 1,6 mm
Price
62.70 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect
Specifications
  • Per:
    Connecting flanged PTFE
    elastic tubing with end fittings and wall thickness 0,8 and 1,6 mm
  • Ø foro:
    1,6 mm
  • Filettatura:
    1/4“ 28G UNF
  • Ø int.tubo:
    1,6 mm
  • Descrizione:
    Couplings with UNF1/4"28G female thread