Orders LinkContactUsLinkComponent
L(-)-Glutatione (forma ridotta)
Glass bottle for solids, 1 g
  CAYM10007461-1
L(-)-Glutatione (forma ridotta)
  CAYM10007461-1
 :  70-18-8
L(-)-Glutatione (forma ridotta)
Glass bottle for solids, 1 g
 :  70-18-8
SFr. 37.00
1 * 1 g
 :  L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
  •  :70-18-8
  •  :C₁₀H₁₇N₃O₆S
  •  :MFCD00065939
  •  :307.33 g/mol
  •  :13,04488
  •  :185…195 °C
  •  :1,475 g/cm³ (20 °C)
  •  :Refrigerata
  •  :200-725-4
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1 g