Single channel pipettes, variable volume
10 - 100 µl
Codice catalogo 613-1360
Nome del fornitore: EDVOTEK
Vai a
Product Details & Documents
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
- Volume easily selected by twisting the top
- Lightweight design and tip ejector makes operation fast and easy
- Tool and instructions included for self-calibration
Specifiche
- Volume:10 - 100 µl