NMR tubes, polyethylene pressure caps
5 mm, giallo
N° de catalogue SCERSP897095-0081
Fournisseur: DWK Life Sciences
NMR tubes, polyethylene pressure caps
N° de catalogue SCERSP897095-0081
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Spécifications
- Colore:giallo
- Lunghezza:5 mm
- Seq:05