Orders LinkContactUsLinkComponent
Homogeniser, Dounce type, KIMBLE®
Homogeniser, Dounce type, KIMBLE®
15 ml
Codice catalogo 432-0252
Nome del fornitore:   DWK Life Sciences
Homogeniser, Dounce type, KIMBLE®
Codice catalogo 432-0252
Nome del fornitore:  DWK Life Sciences
Homogeniser, Dounce type, KIMBLE®
Homogeniser, Dounce type, KIMBLE®
15 ml
Price
SFr. 477.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable
Specifiche
  • Capacità:
    15 ml
  • Seq:
    03