Micro capillaries, disposable, Kimble®
Capillary pipettes, 5 µl, bianco
Catalog # SCERSP71900-5
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
- Calibrated to contain
- Colour-coded
- Marked with single black capacity ring
- Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Packaging: Each cylinder contains 250 pipettes and one pipette device.
Specifications
- Codice colore:bianco
- Capacità:5 µl
- Descrizione:Capillary pipettes
- Seq:01