Universal absorbent pillows
Universal pillow, 20 pillows/pack, 250×250 mm
Codice catalogo MSGA9944
Nome del fornitore: asecos
Vai a
Product Details & Documents
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Optimal for those hard to reach places
- Capacity up to 16 times of its own weight
- High rib and rub resistance
Specifiche
- Descrizione:Universal pillow
- L×L:250×250 mm
- Confezione:20 pillows/pack