Orders LinkContactUsLinkComponent
Density hydrometers, ASTM
1,100 - 1,150 g/ml, 0,001 g/ml, 335 mm
  ALLA3510MC11060MQP
Density hydrometers, ASTM
  ALLA3510MC11060MQP
Density hydrometers, ASTM
1,100 - 1,150 g/ml, 0,001 g/ml, 335 mm
SFr. 60.20
1 * 1 ST
  • Lunghezza:
    335 mm
  • Seq:
    10
  • Divisione:
    0,001 g/ml
  • Range di misura:
    1,100 - 1,150 g/ml